missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cefalonium Dihydrate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Cefalonium Dihydrate |
| CAS | 1385046-35-4 |
| Analyte or Component Names | Cefalonium Dihydrate |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C20 H18 N4 O5 S2 . 2 H2 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4-Carbamoyl-1-[[2-carboxy-8-oxo-7-[2-(2-thienyl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]pyridinium hydroxide, inner salt, 4-(Aminocarbonyl)-1-[[(6R,7R)-2-carboxy-8-oxo-7-[(2-thienylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]pyridinium, inner salt, (6R-trans)-4-(Aminocarbonyl)-1-[[2-carboxy-8-oxo-7-[(2-thienylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]pyridinium hydroxide, inner salt, 4-Carbamoyl-1-[[2-carboxy-8-oxo-7-[2-(2-thienyl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]pyridinium hydroxide, inner salt, 3-(4-Carbamoylpyridylmethyl)-8-oxo-7-[alpha-(thien-2-yl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-(alpha-Thienylacetamido)-3-(4-carbamoylpyridinomethyl)-3-cephem-4-carboxylic acid, Cefalonium, Cepalonium, Cephalonium, Cepravin DC, Cepravin Dry Cow, (6R-trans)-4-(Aminocarbonyl)-1-[[2-carboxy-8-oxo-7-[(2-thienylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]pyridinium , inner salt |
| InChI Formula | InChI=1S/C20H18N4O5S2.2H2O/c21-17(26)11-3-5-23(6-4-11)9-12-10-31-19-15(18(27)24(19)16(12)20(28)29)22-14(25)8-13-2-1-7-30-13;;/h1-7,15,19H,8-10H2,(H3-,21,22,25,26,28,29);2*1H2/t15-,19-;;/m1../s1 |
| Solution Type | Neat |
| SMILES | O.O.NC(=O)c1cc[n+](CC2=C(N3[C@H](SC2)[C@H](NC(=O)Cc4cccs4)C3=O)C(=O)[O-])cc1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?