missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Celiprolol Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal
Description
Celiprolol Hydrochloride 1.0 mg/ml in Methanol (as free base)
Specifications
Specifications
| Chemical Name or Material | Celiprolol Hydrochloride |
| CAS | 57470-78-7 |
| Analyte or Component Names | Celiprolol Hydrochloride |
| Shipping Condition | Ice Pack (-20°C) |
| Molecular Formula | C20 H33 N3 O4 . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Celiprolol Hydrochloride, Celectol, Corliprol, NSC 324509, REV 5320 A, Urea, N'-[3-acetyl-4-[3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]phenyl]-N,N-diethyl-, monohydrochloride (9 CI), Urea, N'-[3-acetyl-4-[3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]phenyl]-N,N-diethyl-, monohydrochloride (9 CI), Urea, N'-[3-acetyl-4-[3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]phenyl]-N,N-diethyl-, monohydrochloride (9 CI) |
| InChI Formula | InChI=1 S/C20H33N3O4.ClH/c1-7-23(8-2)19(26)22-15-9-10-18(17(11-15)14(3)24)27-13-16(25)12-21-20(4,5)6;/h9-11,16,21,25 H,7-8,12-13H2,1-6H3,(H,22,26);1 H |
| SMILES | Cl.CCN(CC)C(=O)Nc1ccc(OCC(O)CNC(C)(C)C)c(c1)C(=O)C |
| IUPAC Name | 3-[3-acetyl-4-[3-(tert-butylamino)-2-hydroxypropoxy]phenyl]-1,1-diethylurea;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?