missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CEP63 Polyclonal antibody specifically detects CEP63 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | CEP63 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:200-1:2000 |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | centrosomal protein 63kDa, centrosomal protein of 63 kDa, centrosome protein CEP63, Cep63, FLJ13386, MGC78416 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human CEP63 (NP_001035859.1). MEALLEGIQNRGHGGGFLTSCEAELQELMKQIDIMVAHKKSEWEGRTHALETCLKIREQELKSLRSQLDVTHKEVGMLHQQVEEHEKIKQEMTMEYKQELKKLHEELCILKRSYEKLQKKQMREFRGNTKNHREDRSEIERLTAKIEEFRQKSLDWEKQRLIYQQQVSSLEAQRKALAEQSEIIQAQLVNRKQKLESVELSSQSEIQHLSSKLERANDTICANELEIERLTMRVNDLVGTSMTVLQEQQQKEEKLRESEKLLEALQEEKRELKAALQSQENLIHEARIQKEKLQEKVKAT |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?