missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cetirizine.2HCl, Lipomed
Description
Cetirizine.2HCl
Specifications
Specifications
| Chemical Name or Material | Cetirizine Dihydrochloride |
| CAS | 83881-52-1 |
| Analyte or Component Names | Cetirizine Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H25 Cl N2 O3 . 2 Cl H |
| Synonym | Cetirizine Dihydrochloride, Acetic acid, [2-[4-[(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]-, dihydrochloride (9 CI), Alercet, Alerid, Alerlisin, Cesta, Cetirizine hydrochloride, Cetrine, Cetrizet, Cistamine, Formistin, Histazine, Nosemin, P 071, Reactine, Riztec, Ryzen, Sancotec, Selitex, Triz, UCB-P 071, Virlix, Zeran, Zirtec, Zirtek, Zirtin, Zyrlex, Zyrtec, Zyrzine, Cetiriz, Cetirizine dihydrochloride, Cetrak, Epirizine, Histazine-1, Tomazine |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.Cl.OC(=O)COCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3 |
| IUPAC Name | 2-[2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetic acid;dihydrochloride |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?