missing translation for 'onlineSavingsMsg'
Learn More

Actin (Muscle Specific) Antibody (MSA/8949R), R&D Systems™

Product Code. 18601565 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18601565 20 μg 20µg
18651144 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18601565 Supplier Novus Biologicals Supplier No. NBP32379920ug
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody

Actin (Muscle Specific) Monoclonal antibody specifically detects Actin (Muscle Specific) in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Actin (Muscle Specific)
Applications ELISA
Classification Monoclonal
Clone MSA/8949R
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation 10 mM PBS with 0.05% BSA
Gene Alias ACTA, ACTC, actin, alpha 2, smooth muscle, aorta, actin, alpha cardiac muscle 1, actin, alpha skeletal muscle, actin, aortic smooth muscle, ACTSA, alpha-cardiac actin, ASD5, ASMA, Cell growth-inhibiting gene 46 protein, CFTD, CFTD1, CFTDM, CMD1R, CMH11, MPFD, NEM1, NEM2, NEM3, nemaline myopathy type 3
Host Species Rabbit
Immunogen Recombinant protein corresponding to human muscle actin
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Developmental Biology, Extracellular Matrix, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 58
Target Species Human
Content And Storage Store at 4°C.
Product Type Antibody
Form Purified
Isotype IgG κ
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.