missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHD7 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-77393SS
This item is not returnable.
View return policy
Description
CHD7 Polyclonal specifically detects CHD7 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CHD7 | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| ATP-dependent helicase CHD7, CHD-7, chromodomain helicase DNA binding protein 7, chromodomain helicase DNA binding protein 7 isoform CRA_e, chromodomain-helicase-DNA-binding protein 7, EC 3.6.1, EC 3.6.4.12, FLJ20357, FLJ20361, IS3, KIAA1416KAL5 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and mouse. Immunogen has 85% identity to chicken Chd7. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.27 mg/mL | |
| Western Blot 1:2500, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:250 | |
| Q9P2D1, A2AJK6 | |
| CHD7 | |
| A genomic peptide made to an N-terminal region of the human Chd7 protein (within residues 25-200). [Swiss-Prot Q9P2D1] | |
| 0.025 mL | |
| Transcription Factors and Regulators | |
| 55636 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction