missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHD7 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
231.00 € - 544.00 €
Specifications
| Antigen | CHD7 |
|---|---|
| Concentration | 1.27 mg/mL |
| Dilution | Western Blot 1:2500, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:250 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18278215
|
Novus Biologicals
NBP1-77393SS |
0.025 mL |
231.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18729303
|
Novus Biologicals
NBP1-77393 |
544.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
CHD7 Polyclonal specifically detects CHD7 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| CHD7 | |
| Western Blot 1:2500, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:250 | |
| Polyclonal | |
| Rabbit | |
| Transcription Factors and Regulators | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| ATP-dependent helicase CHD7, CHD-7, chromodomain helicase DNA binding protein 7, chromodomain helicase DNA binding protein 7 isoform CRA_e, chromodomain-helicase-DNA-binding protein 7, EC 3.6.1, EC 3.6.4.12, FLJ20357, FLJ20361, IS3, KIAA1416KAL5 | |
| CHD7 | |
| IgG | |
| Affinity Purified | |
| Human and mouse. Immunogen has 85% identity to chicken Chd7. |
| 1.27 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Q9P2D1, A2AJK6 | |
| 55636 | |
| A genomic peptide made to an N-terminal region of the human Chd7 protein (within residues 25-200). [Swiss-Prot Q9P2D1] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title