missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chikungunya Virus Antibody (3E7b), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-53118-0.025mg
This item is not returnable.
View return policy
Description
Chikungunya Virus Monoclonal specifically detects Chikungunya Virus in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Chikungunya virus | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Functional | |
| Chikungunya | |
| Live chikungunya virus | |
| 0.025 mg | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| Both Anti-CHIKV [3E7b] and Anti-CHIKV [8A2c] bind strongly to native CHIKV surface and potently neutralize CHIKV replication. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgM |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Functional Assay | |
| 3E7b | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Mouse | |
| Thiophilic Chromatography | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction