missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chikungunya Virus Antibody (3E7b), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
220.00 € - 446.00 €
Specifications
| Antigen | Chikungunya virus |
|---|---|
| Clone | 3E7b |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Functional |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Functional Assay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18220830
|
Novus Biologicals
NBP2-53118 |
0.1 mg |
446.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18644818
|
Novus Biologicals
NBP2-53118-0.025mg |
0.025 mg |
220.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chikungunya Virus Monoclonal specifically detects Chikungunya Virus in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| Chikungunya virus | |
| 1.0 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Functional Assay | |
| Unconjugated | |
| Mouse | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| PBS with 0.02% Sodium Azide | |
| Live chikungunya virus | |
| Primary | |
| Both Anti-CHIKV [3E7b] and Anti-CHIKV [8A2c] bind strongly to native CHIKV surface and potently neutralize CHIKV replication. |
| 3E7b | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Functional | |
| Monoclonal | |
| Purified | |
| RUO | |
| Virus | |
| Chikungunya | |
| IgM | |
| Thiophilic Chromatography |
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit