missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chikungunya Virus Antibody (6A11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
214.00 € - 512.00 €
Specifications
| Antigen | Chikungunya virus |
|---|---|
| Clone | 6A11 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry, Immunocytochemistry/Immunofluorescence |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18256300
|
Novus Biologicals
NBP2-53111 |
0.1 mg |
512.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18637548
|
Novus Biologicals
NBP2-53111-0.025mg |
0.025 mg |
214.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chikungunya Virus Monoclonal specifically detects Chikungunya Virus in Virus samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence.Specifications
| Chikungunya virus | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| PBS with 0.02% Sodium Azide | |
| Live Chikungunya Virus | |
| Primary |
| 6A11 | |
| Western Blot 1:100 - 1:2000, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| Virus | |
| Chikungunya | |
| IgG2b | |
| Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title