missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloramphenicol Antibody - BSA Free, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NB100-62457-0.1ML
This item is not returnable.
View return policy
Description
Chloramphenicol Polyclonal specifically detects Chloramphenicol in Non-species specific samples. It is validated for ELISA.
Specifications
| Chloramphenicol | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.09% Sodium Azide | |
| Chloramphenicol base conjugated via its amino group to KLH using phenylenediisothiocyanate | |
| 0.1 mL | |
| Primary | |
| Non-species specific | |
| Purified |
| ELISA | |
| 5.0 mg/mL | |
| ELISA 1:100-1:2000 | |
| Sheep | |
| IgG purified | |
| RUO | |
| This product recognises chloramphenicol, a broad-spectrum antimicrobial that inhibits protein synthesis. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction