missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloramphenicol Antibody - BSA Free, Novus Biologicals™
Sheep Polyclonal Antibody
197.00 € - 456.00 €
Specifications
| Antigen | Chloramphenicol |
|---|---|
| Concentration | 5.0 mg/mL |
| Dilution | ELISA 1:100-1:2000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18093086
|
Novus Biologicals
NB100-62457-0.1ML |
0.1 mL |
197.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18770254
|
Novus Biologicals
NB100-62457 |
456.00 €
0.50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Chloramphenicol Polyclonal specifically detects Chloramphenicol in Non-species specific samples. It is validated for ELISA.Specifications
| Chloramphenicol | |
| ELISA 1:100-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 0.09% Sodium Azide | |
| IgG | |
| IgG purified | |
| This product recognises chloramphenicol, a broad-spectrum antimicrobial that inhibits protein synthesis. |
| 5.0 mg/mL | |
| ELISA | |
| Unconjugated | |
| Sheep | |
| Non-species specific | |
| Chloramphenicol base conjugated via its amino group to KLH using phenylenediisothiocyanate | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title