missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlormadinone Acetate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Chlormadinone acetate |
| CAS | 302-22-7 |
| Analyte or Component Names | Chlormadinone acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H29 Cl O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Pregna-4,6-diene-3,20-dione, 6-chloro-17-hydroxy-, acetate (6CI,7CI,8CI), 17-Acetoxy-6-chloro-6-dehydroprogesterone, 17-Acetoxy-6-chloropregna-4,6-diene-3,20-dione, 6-Chloro-17-acetoxy-4,6-pregnadiene-3,20-dione, 6-Chloro-17-hydroxypregna-4,6-diene-3,20-dione acetate, 6-Chloro-6,7-dehydro-17-acetoxyprogesterone, 6-Chloro-6-dehydro-17-acetoxyprogesterone, Ay 13390-6, Bellissima, Bovisynchron, CAP, Cero, Chlormadinon acetate, Chlormadinone acetate, Chronosyn, Clordion, Cyclonorm, Fertiletten, Gestafortin, Lormin, Luteran, Lutinyl, Lutoral, Lutoral (Syntex), Matrol, Menstridyl, NSC 92338, Natrol, Normenon, Prostal, RS 1280, Retex, Skedule TM, Synchrosyn, Synchrosyn P, Traslan, Verton, 17-(Acetyloxy)-6-chloropregna-4,6-diene-3,20-dione, 17alpha-Acetoxy-6-chloro-4,6-pregnadiene-3,20-dione, 17alpha-Acetoxy-6-chloro-6,7-dehydroprogesterone, 6-Chloro-17alpha-acetoxy-4,6-pregnadiene-3,20-dione, 6-Chloro-17alpha-hydroxy-4,6-pregnadiene-3,20-dione 17-acetate, 6-Chloro-17alpha-hydroxy-Delta6-progesterone acetate, 6-Chloro-17alpha-hydroxypregna-4,6-diene-3,20-dione acetate, 6-Chloro-6-dehydro-17alpha-acetoxyprogesterone, 6-Chloro-pregna-4,6-dien-17alpha-ol-3,20-dione acetate, 6-Chloro-Delta4,6-pregnadien-17alpha-ol-3,20-dione 17-acetate, 6-Chloro-Delta6-17-acetoxyprogesterone, 6-Chloro-Delta6-[17alpha]acetoxyprogesterone, 6-Chloro-Delta6-dehydro-17-acetoxyprogesterone, 6-Dehydro-6-chloro-17alpha-acetoxyprogesterone |
| InChI Formula | InChI=1S/C23H29ClO4/c1-13(25)23(28-14(2)26)10-7-18-16-12-20(24)19-11-15(27)5-8-21(19,3)17(16)6-9-22(18,23)4/h11-12,16-18H,5-10H2,1-4H3/t16-,17+,18+,21-,22+,23+/m1/s1 |
| Solution Type | Neat |
| SMILES | CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C=C(Cl)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(=O)C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?