missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloropyrilene Citrate, LoGiCal
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
Chloropyrilene Citrate
Specifications
Specifications
| Chemical Name or Material | Chloropyrilene Citrate |
| CAS | 148-64-1 |
| Analyte or Component Names | Chloropyrilene Citrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H18 Cl N3 S . C6 H8 O7 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1,2-Ethanediamine, N1-[(5-chloro-2-thienyl)methyl]-N2,N2-dimethyl-N1-2-pyridinyl-, 2-hydroxy-1,2,3-propanetricarboxylate (1:1), 1,2-Ethanediamine, N-[(5-chloro-2-thienyl)methyl]-N',N'-dimethyl-N-2-pyridinyl-, 2-hydroxy-1,2,3-propanetricarboxylate (1:1), Pyridine, 2-[(5-chloro-2-thenyl)[2-(dimethylamino)ethyl]amino]-, citrate (1:1), 2-[(5-Chloro-2-thenyl)(2-dimethylaminoethyl)amino]pyridine citrate, 2-[(5-Chloro-2-thenyl)[2-(dimethylamino)ethyl]amino]pyridine Dihydrogen Citrate, Chloropyrilenium citrate, Chlorotene citrate, Chlorothen citrate, Chlorothenium citrate, Tagafen |
| InChI Formula | InChI=1 S/C14H18ClN3S.C6H8O7/c1-17(2)9-10-18(14-5-3-4-8-16-14)11-12-6-7-13(15)19-12;7-3(8)1-6(13,5(11)12)2-4(9)10/h3-8 H,9-11H2,1-2H3;13 H,1-2H2,(H,7,8)(H,9,10)(H,11,12) |
| SMILES | CN(C)CCN(Cc1ccc(Cl)s1)c2ccccn2.OC(=O)CC(O)(CC(=O)O)C(=O)O |
| IUPAC Name | N'-[(5-chlorothiophen-2-yl)methyl]-N,N-dimethyl-N'-pyridin-2-ylethane-1,2-diamine;2-hydroxypropane-1,2,3-tricarboxylic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction