missing translation for 'onlineSavingsMsg'
Learn More

Chlorpromazine hydrochloride, Dr. Ehrenstorfer

Product Code. 30316277
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316277 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316277 Supplier Dr. Ehrenstorfer Supplier No. DREC11575000100MG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlorpromazine Hydrochloride
CAS 69-09-0
Analyte or Component Names Chlorpromazine Hydrochloride
Shipping Condition +20°C
Molecular Formula C17 H19 Cl N2 S . Cl H
Certifications/Compliance ISO 17034
Synonym Chlorpromazine Hydrochloride, 10H-Phenothiazine-10-propanamine, 2-chloro-N,N-dimethyl-, monohydrochloride (9CI), Phenothiazine, 2-chloro-10-[3-(dimethylamino)propyl]-, monohydrochloride (8CI), 10-(3-Dimethylaminopropyl)-2-chlorophenothiazine monohydrochloride, 2-Chloro-10-(3-dimethylaminopropyl)phenothiazine monohydrochloride, Aminazin hydrochloride, Aminazin monohydrochloride, Ampliactil monohydrochloride, CPZ, Chloractil, Chlorazin, Chloropromazine hydrochloride, Chloropromazine monohydrochloride, Chlorpromazine monohydrochloride, Chlorpromazinium chloride, Hebanil, Hibanil, Hibernal, Klorproman, Klorpromex, Largactil, Largactil monohydrochloride, Largaktyl, Marazine, Megatil, Neurazine, Norcozine, Plegomazin, Promacid, Promapar, Propaphen, Propaphenin hydrochloride, Sonazine, Taroctyl, Thorazine hydrochloride, Torazina, Tranzene
InChI Formula InChI=1S/C17H19ClN2S.ClH/c1-19(2)10-5-11-20-14-6-3-4-7-16(14)21-17-9-8-13(18)12-15(17)20;/h3-4,6-9,12H,5,10-11H2,1-2H3;1H
SMILES Cl.CN(C)CCCN1c2ccccc2Sc3ccc(Cl)cc13
IUPAC Name 3-(2-chlorophenothiazin-10-yl)-N,N-dimethylpropan-1-amine;hydrochloride
Molecular Weight (g/mol) 355.33
Quantity 100 mg
Formula Weight 354.0724 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.