missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorpromazine Hydrochloride, Mikromol™
Description
Chlorpromazine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Chlorpromazine Hydrochloride |
| CAS | 69-09-0 |
| Analyte or Component Names | Chlorpromazine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H19 Cl N2 S . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Chlorpromazine Hydrochloride, 10H-Phenothiazine-10-propanamine, 2-chloro-N,N-dimethyl-, monohydrochloride (9CI), Phenothiazine, 2-chloro-10-[3-(dimethylamino)propyl]-, monohydrochloride (8CI), 10-(3-Dimethylaminopropyl)-2-chlorophenothiazine monohydrochloride, 2-Chloro-10-(3-dimethylaminopropyl)phenothiazine monohydrochloride, Aminazin hydrochloride, Aminazin monohydrochloride, Ampliactil monohydrochloride, CPZ, Chloractil, Chlorazin, Chloropromazine hydrochloride, Chloropromazine monohydrochloride, Chlorpromazine monohydrochloride, Chlorpromazinium chloride, Hebanil, Hibanil, Hibernal, Klorproman, Klorpromex, Largactil, Largactil monohydrochloride, Largaktyl, Marazine, Megatil, Neurazine, Norcozine, Plegomazin, Promacid, Promapar, Propaphen, Propaphenin hydrochloride, Sonazine, Taroctyl, Thorazine hydrochloride, Torazina, Tranzene |
| InChI Formula | InChI=1S/C17H19ClN2S.ClH/c1-19(2)10-5-11-20-14-6-3-4-7-16(14)21-17-9-8-13(18)12-15(17)20;/h3-4,6-9,12H,5,10-11H2,1-2H3;1H |
| SMILES | Cl.CN(C)CCCN1c2ccccc2Sc3ccc(Cl)cc13 |
| IUPAC Name | 3-(2-chlorophenothiazin-10-yl)-N,N-dimethylpropan-1-amine;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?