missing translation for 'onlineSavingsMsg'
Learn More

Chlortetracycline Hydrochloride, Mikromol™

Product Code. 17931506
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17931506 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17931506 Supplier Mikromol Supplier No. MM0607.00
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Chlortetracycline Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlortetracycline Hydrochloride
CAS 64-72-2
Analyte or Component Names Chlortetracycline Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C22 H23 Cl N2 O8 . Cl H
Certifications/Compliance ISO 9001
Synonym Chlortetracycline Hydrochloride, (4S,4aS,5aS,6S,12aS)-7-Chloro-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide Monohydrochloride, [4S-(4alpha,4aalpha,5aalpha,6beta,12aalpha)]-7-Chloro-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide Monohydrochloride, 7-Chlorotetracycline hydrochloride, 7-Chlorotetracycline monohydrochloride, Aureociclina, Aureocycline, Aureomycin hydrochloride, Aureomycin monohydrochloride, Aureovit 12C80, Aurofac 100, Biomycin hydrochloride, Chlortetracycline hydrochloride, Chlortetracyclinium chloride, Fermycin Soluble, Isphamycin, NSC 13252
InChI Formula InChI=1S/C22H23ClN2O8.ClH/c1-21(32)7-6-8-15(25(2)3)17(28)13(20(24)31)19(30)22(8,33)18(29)11(7)16(27)12-10(26)5-4-9(23)14(12)21;/h4-5,7-8,15,26,28-29,32-33H,6H2,1-3H3,(H2,24,31);1H/t7-,8-,15-,21-,22-;/m0./s1
SMILES Cl.CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N)C(=O)c4c(O)ccc(Cl)c4[C@@]3(C)O
IUPAC Name (4S,4aS,5aS,6S,12aS)-7-chloro-4-(dimethylamino)-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
Molecular Weight (g/mol) 515.34
Quantity 500 mg
Formula Weight 514.091 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.