missing translation for 'onlineSavingsMsg'
Learn More

Chlorthalidone, Dr. Ehrenstorfer

Product Code. 30324324
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30324324 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324324 Supplier Dr. Ehrenstorfer Supplier No. DREC11615000
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlortalidone
CAS 77-36-1
Analyte or Component Names Chlortalidone
Shipping Condition +20°C
Molecular Formula C14 H11 Cl N2 O4 S
Certifications/Compliance ISO/IEC 17025
Synonym Benzenesulfonamide, 2-chloro-5-(2,3-dihydro-1-hydroxy-3-oxo-1H-isoindol-1-yl)-, Benzenesulfonamide, 2-chloro-5-(1-hydroxy-3-oxo-1-isoindolinyl)- (6CI,8CI), 2-Chloro-5-(2,3-dihydro-1-hydroxy-3-oxo-1H-isoindol-1-yl)benzenesulfonamide, 1-Oxo-3-(3-sulfamyl-4-chlorophenyl)-3-hydroxy-isoindolinum, 1-Oxo-3-(3-sulfamyl-4-chlorophenyl)-3-hydroxyisoindoline, 1-keto-3-(3'-Sulfamyl-4'-chlorophenyl)-3-hydroxyisoindoline, 2-Chloro-5-(1-hydroxy-3-oxo-1-isoindolinyl)benzenesulfonamide, 2-Chloro-5-(3-hydroxy-1-oxoisoindol-3-yl)benzenesulfonamide, 3-(4'-Chloro-3'-sulfamoylphenyl)-3-hydroxyphthalimidine, 3-Hydroxy-3-[4-chloro-3-sulfamylphenyl]phthalimidine, Chlorothalidone, Chlortalidone, Chlorthalidon, Chlorthalidone, Demi-Regroton, G 33182, Higroton, Higrotona, Hygroton, Igroton, Isoren, NSC 69200, Natriuran, Oradil, Oxodolin, Phthalamudine, Racemic chlorthalidone, Renon, Saluretin, Thalitone, ZINC 00020253, Zambesil, (+/-)-Chlorthalidone, (+/-)-Hygroton
InChI Formula InChI=1S/C14H11ClN2O4S/c15-11-6-5-8(7-12(11)22(16,20)21)14(19)10-4-2-1-3-9(10)13(18)17-14/h1-7,19H,(H,17,18)(H2,16,20,21)
SMILES NS(=O)(=O)c1cc(ccc1Cl)C2(O)NC(=O)c3ccccc23
IUPAC Name 2-chloro-5-(1-hydroxy-3-oxo-2H-isoindol-1-yl)benzenesulfonamide
Molecular Weight (g/mol) 338.77
Quantity 50 mg
Formula Weight 338.0128 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.