missing translation for 'onlineSavingsMsg'
Learn More

CHMP1B Antibody - BSA Free, Novus Biologicals™

Product Code. 30558806 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30558806 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30558806 Supplier Novus Biologicals Supplier No. NBP347623

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CHMP1B Polyclonal antibody specifically detects CHMP1B in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHMP1B
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry 1:50 - 1:100
Formulation PBS, 50% Glycerol, pH (7.3)
Gene Alias C10orf2, c18orf2, charged multivesicular body protein 1b, CHMP1.5, CHMP1b, chromatin modifying protein 1B, Chromatin-modifying protein 1b, hVps46-2, vacuolar protein sorting 46-2, Vacuolar protein sorting-associated protein 46-2, Vps46-2, Vps46B
Host Species Rabbit
Immunogen chromatin modifying protein 1B. (Uniprot# Q7LBR1)
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 57132
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.