missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chorionic Gonadotropin beta Chain (hCG beta) Antibody (INN-hCG-53) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-63167-0.05MG
This item is not returnable.
View return policy
Description
Chorionic Gonadotropin beta Chain (hCG beta) Monoclonal specifically detects Chorionic Gonadotropin beta Chain (hCG beta) in Human samples. It is validated for Western Blot, ELISA.
Specifications
| hCG holo C2 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| P01233 | |
| CGB3 | |
| Made to Human hCG holo C2 | |
| 0.05 mg | |
| Cancer | |
| 1082 | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| INN-hCG-53 | |
| Unconjugated | |
| PBS (pH 7.2) with 0.09% Sodium Azide | |
| choriogonadotropin subunit beta, CGB3, CGB5, CGB7, CGB8, chorionic gonadotropin, beta polypeptide, hCGB | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| NB100-63167 recognizes epitope c2 on hCG holo C2 . It does not recognize free hCG subunits. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction