missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chorionic Gonadotropin beta Chain (hCG beta) Antibody (INN-hCG-53) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
216.00 € - 511.00 €
Specifications
| Antigen | hCG holo C2 |
|---|---|
| Clone | INN-hCG-53 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
| Applications | ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18036234
|
Novus Biologicals
NB100-63167-0.05MG |
0.05 mg |
216.00 €
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18006955
|
Novus Biologicals
NB100-63167 |
0.25 mg |
511.00 €
0.25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chorionic Gonadotropin beta Chain (hCG beta) Monoclonal specifically detects Chorionic Gonadotropin beta Chain (hCG beta) in Human samples. It is validated for Western Blot, ELISA.Specifications
| hCG holo C2 | |
| 1.0 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| P01233 | |
| 1082 | |
| Made to Human hCG holo C2 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| INN-hCG-53 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| choriogonadotropin subunit beta, CGB3, CGB5, CGB7, CGB8, chorionic gonadotropin, beta polypeptide, hCGB | |
| CGB3 | |
| IgG1 | |
| Protein A or G purified | |
| NB100-63167 recognizes epitope c2 on hCG holo C2 . It does not recognize free hCG subunits. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title