missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beskrivning
CIB2 Monoclonal antibody specifically detects CIB2 in Human samples. It is validated for ELISA
Specifikationer
Specifikationer
| Antigen | CIB2 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 12 |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:1000-1:2000 |
| Formulation | 0.2 um filtered solution in PBS |
| Gene Alias | 2810434I23Rik, calcium and integrin binding family member 2, DNA-dependent protein kinase catalytic subunit-interacting protein 2, Kinase-interacting protein 2, KIP 2, KIP2calcium and integrin-binding family member 2 |
| Host Species | Mouse |
| Immunogen | This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CIB2 (Uniprot#: O75838; Met1-Ile187). |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?