missing translation for 'onlineSavingsMsg'
Learn More

Ciprofloxacin Hydrochloride Monohydrate, LoGiCal

Product Code. 30458732
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity unitSize
30458732 10 mg 10mg
1 options
This item is not returnable. View return policy

Product Code. 30458732

Brand: LoGiCal LGCFOR0018.06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ciprofloxacin Hydrochloride Monohydrate
CAS 86393-32-0
Analyte or Component Names Ciprofloxacin Hydrochloride Monohydrate
Shipping Condition Room Temperature
Molecular Formula C17 H18 F N3 O3 . Cl H . H2 O
Certifications/Compliance ISO 9001
Synonym 3-Quinolinecarboxylic acid, 1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-, monohydrochloride (9CI), Adiflox, Alcon Cilox, BAY-o 9867, Bacquinor, Bactiflox, Baflox, Baycip, Belmacina, Bernoflox, Cetraxal, Ciflodal, Ciflox, Cifloxin, Cifran, Cilab, Ciloxan, Cimogal, Ciplox, Ciploxx, Ciplus, Ciprecu, Ciprinol, Cipro, Cipro (pharmaceutical), Cipro optic, Ciprobay, Ciprobay Uro, Ciproben 250, Ciprobid, Ciprobiotic, Ciprocin, Ciprocinol, Ciprodar, Ciprofar, Ciproflox, Ciprofloxacin hydrochloride, Ciprogis, Ciprok, Ciprolin, Ciprolon, Cipromax, Cipromid, Cipromycin, Ciproquinol, Ciprowin, Ciproxan, Ciproxin, Ciproxine, Ciriax, Citopcin, Cixan, Corsacin, Cosflox, Cycin, Cyprobay, Eni, Estecina, Fimoflox, Ipiflox, Kenzoflex, Loxan, Medociprin, Microrgan, Mitroken, NSC 620634, Nivoflox, Opthaflox, Probiox, Proflaxin, Proflox, Proksi 250, Proksi 500, Quindrops, Quinolide, Quintor, Rancif, Ringoran, Roxytal, Septicide, Serviflox, Sipro 250, Sophixin Ofteno, Spitacin, Superocin, Unex, Uniflox, Zoxan, Zumaflox, Enrofloxacin Imp. B (EP), Ciprofloxacin Hydrochloride Monohydrate, Enrofloxacin Imp. B (EP) as Hydrochloride Monohydrate, 1-Cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid monohydrochloride monohydrate, Enrofloxacin Impurity B as Hydrochloride Monohydrate
InChI Formula InChI=1S/C17H18FN3O3.ClH.H2O/c18-13-7-11-14(8-15(13)20-5-3-19-4-6-20)21(10-1-2-10)9-12(16(11)22)17(23)24;;/h7-10,19H,1-6H2,(H,23,24);1H;1H2
Solution Type Neat
SMILES O.Cl.OC(=O)C1=CN(C2CC2)c3cc(N4CCNCC4)c(F)cc3C1=O
IUPAC Name 1-cyclopropyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid;hydrate;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 385.82
Quantity 10 mg
Formula Weight 385.1205 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.