missing translation for 'onlineSavingsMsg'
Learn More

Ciprofloxacin Hydrochloride Monohydrate, Mikromol™

Product Code. 17972606
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17972606 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17972606 Supplier Mikromol Supplier No. MM0018.06
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ciprofloxacin Hydrochloride Monohydrate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ciprofloxacin Hydrochloride Monohydrate
CAS 86393-32-0
Analyte or Component Names Ciprofloxacin Hydrochloride Monohydrate
Shipping Condition Room Temperature
Molecular Formula C17 H18 F N3 O3 . Cl H . H2 O
Certifications/Compliance ISO 17025
Synonym 3-Quinolinecarboxylic acid, 1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-, monohydrochloride (9CI), Adiflox, Alcon Cilox, BAY-o 9867, Bacquinor, Bactiflox, Baflox, Baycip, Belmacina, Bernoflox, Cetraxal, Ciflodal, Ciflox, Cifloxin, Cifran, Cilab, Ciloxan, Cimogal, Ciplox, Ciploxx, Ciplus, Ciprecu, Ciprinol, Cipro, Cipro (pharmaceutical), Cipro optic, Ciprobay, Ciprobay Uro, Ciproben 250, Ciprobid, Ciprobiotic, Ciprocin, Ciprocinol, Ciprodar, Ciprofar, Ciproflox, Ciprofloxacin hydrochloride, Ciprogis, Ciprok, Ciprolin, Ciprolon, Cipromax, Cipromid, Cipromycin, Ciproquinol, Ciprowin, Ciproxan, Ciproxin, Ciproxine, Ciriax, Citopcin, Cixan, Corsacin, Cosflox, Cycin, Cyprobay, Eni, Estecina, Fimoflox, Ipiflox, Kenzoflex, Loxan, Medociprin, Microrgan, Mitroken, NSC 620634, Nivoflox, Opthaflox, Probiox, Proflaxin, Proflox, Proksi 250, Proksi 500, Quindrops, Quinolide, Quintor, Rancif, Ringoran, Roxytal, Septicide, Serviflox, Sipro 250, Sophixin Ofteno, Spitacin, Superocin, Unex, Uniflox, Zoxan, Zumaflox, Enrofloxacin Imp. B (EP), Ciprofloxacin Hydrochloride Monohydrate, Enrofloxacin Imp. B (EP) as Hydrochloride Monohydrate, 1-Cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid monohydrochloride monohydrate, Enrofloxacin Impurity B as Hydrochloride Monohydrate
InChI Formula InChI=1S/C17H18FN3O3.ClH.H2O/c18-13-7-11-14(8-15(13)20-5-3-19-4-6-20)21(10-1-2-10)9-12(16(11)22)17(23)24;;/h7-10,19H,1-6H2,(H,23,24);1H;1H2
SMILES O.Cl.OC(=O)C1=CN(C2CC2)c3cc(N4CCNCC4)c(F)cc3C1=O
IUPAC Name 1-cyclopropyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid;hydrate;hydrochloride
Molecular Weight (g/mol) 385.82
Quantity 500 mg
Formula Weight 385.1205 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.