missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Aminocyclohexanol Hydrochloride, Mikromol

Product Code. 30462148
Change view
Click to view available options
Quantity:
25 mg
4 x 25 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30462148 4 x 25 mg 100mg
30051690 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30462148 Supplier Mikromol Supplier No. MM0005.0700254AMP
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

cis-4-Aminocyclohexanol Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material cis-4-Aminocyclohexanol Hydrochloride
CAS 56239-26-0
Analyte or Component Names cis-4-Aminocyclohexanol Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C6 H13 N O . Cl H
Certifications/Compliance ISO 9001
Synonym cis-4-Aminocyclohexanol Hydrochloride
InChI Formula InChI=1S/C6H13NO.ClH/c7-5-1-3-6(8)4-2-5;/h5-6,8H,1-4,7H2;1H/t5-,6+;
Solution Type Neat
SMILES Cl.N[C@@H]1CC[C@H](O)CC1
IUPAC Name 4-aminocyclohexan-1-ol;hydrochloride
Molecular Weight (g/mol) 151.63
Quantity 4 x 25 mg
Formula Weight 151.0764 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.