missing translation for 'onlineSavingsMsg'
Learn More

CISD2 Antibody (8A0) [mFluor Violet 450 SE], Novus Biologicals Biologicals™

Product Code. 30518759 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30518759 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30518759 Supplier Novus Biologicals Supplier No. NBP342942MFV450

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Mouse Monoclonal Antibody

CISD2 Monoclonal antibody specifically detects CISD2 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Spécification

Antigen CISD2
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 8A0
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias CDGSH iron sulfur domain 2, CDGSH iron-sulfur domain-containing protein 2, CDGSH2, Endoplasmic reticulum intermembrane small protein, ERISWFS2, Miner1Wolfram syndrome 2, mitoNEET related 1, MitoNEET-related 1 protein, NAF-1, Nutrient-deprivation autophagy factor-1, ZCD2CDGSH iron sulfur domain-containing protein 2, zinc finger, CDGSH-type domain 2
Host Species Mouse
Immunogen CDGSH iron sulfur domain 2 - (Uniprot# Q8N5K1)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 493856
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.