missing translation for 'onlineSavingsMsg'
Learn More

Citalopram N-Oxide, Mikromol™

Product Code. 30051025
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051025 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051025 Supplier Mikromol Supplier No. MM0242.10
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Citalopram N-Oxide
TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Citalopram N-Oxide
CAS 63284-72-0
Analyte or Component Names Citalopram N-Oxide
Shipping Condition Ice pack (-20°C)
Molecular Formula C20 H21 F N2 O2
Certifications/Compliance ISO 9001
Synonym 5-Isobenzofurancarbonitrile, 1-[3-(dimethyloxidoamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-, 5-Isobenzofurancarbonitrile, 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-, N1-oxide, 1-[3-(Dimethyloxidoamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile, Citalopram N-oxide, Lu 11-305, Citalopram Related Compound E, Escitalopram Oxalate Impurity H, Escitalopram Impurity H
InChI Formula InChI=1S/C20H21FN2O2/c1-23(2,24)11-3-10-20(17-5-7-18(21)8-6-17)19-9-4-15(13-22)12-16(19)14-25-20/h4-9,12H,3,10-11,14H2,1-2H3
SMILES C[N+](C)([O-])CCCC1(OCc2cc(ccc12)C#N)c3ccc(F)cc3
IUPAC Name 3-[5-cyano-1-(4-fluorophenyl)-3H-2-benzofuran-1-yl]-N,N-dimethylpropan-1-amine oxide
Molecular Weight (g/mol) 340.39
Quantity 100 mg
Formula Weight 340.1587 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.