missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cited-2 Antibody (JA22) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 12 publications
Brand: Novus Biologicals NB100-136SS
This item is not returnable.
View return policy
Description
Cited-2 Monoclonal specifically detects Cited-2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Cited-2 | |
| Monoclonal | |
| 1.0 mg/ml | |
| Western Blot 2ug/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 19319572) | |
| Q99967 | |
| CITED2 | |
| Human CITED 2 protein [UniProt# Q99967] | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Ascites |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| JA22 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain2,cbp/p300-interacting transactivator 2, MRG-1, MRG1melanocyte-specific gene 1-related gene 1, MSG1-related gene 1, MSG-related protein 1, P35srj | |
| Mouse | |
| 29 kDa | |
| 0.025 mL | |
| Cancer, Chromatin Research, HIF Target Genes, Hypoxia | |
| 10370 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction