missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cited-2 Antibody (JA22) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 12 publications
226.00 € - 517.00 €
Specifications
| Antigen | Cited-2 |
|---|---|
| Clone | JA22 |
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 2ug/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 19319572) |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18084365
|
Novus Biologicals
NB100-136SS |
0.025 mL |
226.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18798823
|
Novus Biologicals
NB100-136 |
517.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Cited-2 Monoclonal specifically detects Cited-2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Cited-2 | |
| 1.0 mg/ml | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Chromatin Research, HIF Target Genes, Hypoxia | |
| PBS with 0.02% Sodium Azide | |
| Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain2,cbp/p300-interacting transactivator 2, MRG-1, MRG1melanocyte-specific gene 1-related gene 1, MSG1-related gene 1, MSG-related protein 1, P35srj | |
| CITED2 | |
| IgG1 κ | |
| Protein A or G purified | |
| 29 kDa |
| JA22 | |
| Western Blot 2ug/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 19319572) | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Mouse | |
| Q99967 | |
| 10370 | |
| Human CITED 2 protein [UniProt# Q99967] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title