missing translation for 'onlineSavingsMsg'
Learn More

Citicoline Sodium, Mikromol™

Product Code. 17949414
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17949414 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17949414 Supplier Mikromol Supplier No. MM3913.000250
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Citicoline Sodium
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Citicoline sodium
CAS 33818-15-4
Analyte or Component Names Citicoline sodium
Shipping Condition Room Temperature
Molecular Formula C14 H25 N4 O11 P2 . Na
Certifications/Compliance ISO 17025
Synonym Cytidine 5'-(trihydrogen diphosphate), P'-[2-(trimethylammonio)ethyl] ester, inner salt, sodium salt (1:1), Choline, hydroxide, monoester with cytidine 5'-(trihydrogen pyrophosphate), inner salt, monosodium salt (8CI), Cytidine 5'-(trihydrogen diphosphate), P'-[2-(trimethylammonio)ethyl] ester, hydroxide, inner salt, monosodium salt, Cytidine 5'-(trihydrogen diphosphate), P'-[2-(trimethylammonio)ethyl] ester, inner salt, monosodium salt (9CI), Acticolin, Brassel, Cebroton, CerAxon, Cidifos, Citicoline sodium, Flussorex, Gerolin, IP 302 sodium, Logan, Neurotron, Sinkron, Strolin
InChI Formula InChI=1S/C14H26N4O11P2.Na/c1-18(2,3)6-7-26-30(22,23)29-31(24,25)27-8-9-11(19)12(20)13(28-9)17-5-4-10(15)16-14(17)21;/h4-5,9,11-13,19-20H,6-8H2,1-3H3,(H3-,15,16,21,22,23,24,25);/q;+1/p-1/t9-,11-,12-,13-;/m1./s1
SMILES [Na+].C[N+](C)(C)CCOP(=O)([O-])OP(=O)([O-])OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=CC(=NC2=O)N
IUPAC Name sodium;[[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] 2-(trimethylazaniumyl)ethyl phosphate
Molecular Weight (g/mol) 510.31
Quantity 250 mg
Formula Weight 510.0893 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.