missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Monohydrate, Mikromol™

Product Code. 17912626
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17912626 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17912626 Supplier Mikromol Supplier No. MM0575.00
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Citric Acid Monohydrate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Citric acid monohydrate
CAS 5949-29-1
Analyte or Component Names Citric acid monohydrate
Shipping Condition Room Temperature
Molecular Formula C6 H8 O7 . H2 O
Certifications/Compliance ISO 17034
Synonym Citric Acid Monohydrate, 2-Hydroxypropane-1,2,3-tricarboxylic acid monohydrate
InChI Formula InChI=1S/C6H8O7.H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);1H2
SMILES O.OC(=O)CC(O)(CC(=O)O)C(=O)O
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
Molecular Weight (g/mol) 210.14
Quantity 500 mg
Formula Weight 210.0376 g/mol
Show More Show Less
compliance-icons
Product Identifier
  • Citric Acid Monohydrate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.