missing translation for 'onlineSavingsMsg'
Learn More

Claudin-8 Antibody (474210), Allophycocyanin, R&D Systems™

Product Code. 18613075 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18613075 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18613075 Supplier Novus Biologicals Supplier No. FAB5275A
  • ONLINE EXCLUSIVE PRICE
    728.45 €
    627.90 € / 0.1mL
    Save 100.55 €
    13% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

Claudin-8 Monoclonal specifically detects Claudin-8 in Human samples. It is validated for CyTOF, Flow Cytometry, Flow Cytometry (Intracellular Staining).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Claudin-8
Applications CyTOF, Flow Cytometry, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 474210
Conjugate APC
Dilution Flow Cytometry, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias claudin 8, human CLDN8 gene for claudin-810claudin-8
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human Claudin-8, Met1-Val225 (Ser151Pro), Accession # P56748
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9073
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.