missing translation for 'onlineSavingsMsg'
Learn More

Clevudine, Thermo Scientific™

Product Code. 17525575 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17525575 250 mg 250mg
17515575 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17525575 Supplier Thermo Scientific Acros Supplier No. 469122500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 163252-36-6
Molecular Formula C10H13FN2O5
Molecular Weight (g/mol) 260.22
MDL Number MFCD00935785
InChI Key GBBJCSTXCAQSSJ-XQXXSGGOSA-N
IUPAC Name 1-[(2S,3R,4S,5S)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES CC1=CN([C@H]2O[C@@H](CO)[C@H](O)[C@H]2F)C(=O)NC1=O

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Powder
Color White to Light Yellow
Quantity 250 mg
Chemical Name or Material Clevudine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.