missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clindamycin Hydrochloride, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Clindamycin hydrochloride |
| CAS | 21462-39-5 |
| Analyte or Component Names | Clindamycin hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H33 Cl N2 O5 S . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Clindamycin Hydrochloride, 7(S)-Chloro-7-deoxylincomycin hydrochloride, 7-Chloro-7-deoxylincomycin hydrochloride, Antirobe vet., Cleocin hydrochloride, Clindamycin hydrochloride, Clindavet, Clindimycin hydrochloride, Clindomycin hydrochloride, Dalacina, Methyl 7-Chloro-6,7,8-trideoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-L-threo-alpha-D-galacto-octopyranoside hydrochloride (1:1), trans-alpha-Methyl 7-Chloro-6,7,8-trideoxy-6-(1-methyl-4-propyl-L-2-pyrrolidinecarboxamido)-1-thio-L-threo-alpha-D-galacto-octopyranoside monohydrochloride, (2S-trans)-Methyl 7-Chloro-6,7,8-trideoxy-6-[[(1-methyl-4-propyl-2-pyrrolidinyl)carbonyl]amino]-1-thio-L-threo-alpha-D-galacto-octopyranoside monohydrochloride, Methyl 7-Chloro-6,7,8-trideoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-L-threo-alpha-D-galacto-octopyranoside monohydrochloride |
| InChI Formula | InChI=1S/C18H33ClN2O5S.ClH/c1-5-6-10-7-11(21(3)8-10)17(25)20-12(9(2)19)16-14(23)13(22)15(24)18(26-16)27-4;/h9-16,18,22-24H,5-8H2,1-4H3,(H,20,25);1H/t9-,10+,11-,12+,13-,14+,15+,16+,18+;/m0./s1 |
| Solution Type | Neat |
| SMILES | Cl.CCC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@H](C)Cl)[C@H]2O[C@H](SC)[C@H](O)[C@@H](O)[C@H]2O |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction