missing translation for 'onlineSavingsMsg'
Learn More

Clobetasol Propionate, Mikromol™

Product Code. 18041040
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18041040 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18041040 Supplier Mikromol Supplier No. MM0769.00
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Clobetasol Propionate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Clobetasol Propionate
CAS 25122-46-7
Analyte or Component Names Clobetasol Propionate
Shipping Condition Room Temperature
Molecular Formula C25 H32 Cl F O5
Certifications/Compliance ISO 17034
Synonym Clobetasol Propionate, 21-Chloro-21-deoxybetamethasone 17-propionate, CCl 4725, CGP 9555, Clobederm, Clobesol, Clobetasol 17-propionate, Clobetasol propionate, Clobex, Dermoval, Dermovate, Dermoxin, Dermoxinale, GR 2/925, Karison, Olux, Olux-E, Skin Cap, Temovate, (11beta,16beta)-21-Chloro-9-fluoro-11-hydroxy-16-methyl-17-(1-oxopropoxy)pregna-1,4-diene-3,20-dione, 21-Chloro-9-fluoro-11beta,17-dihydroxy-16beta-methylpregna-1,4-diene-3,20-dione 17-propionate, AH 22175X, CCI 4725, CCI18747, CCI6990, Clobetasol-17-propionate, CCI1822, 21-desoxybetamethasone-17-propionate, 16-ene, USP Clobetasol propionate Related Compound A
InChI Formula InChI=1S/C25H32ClFO5/c1-5-21(31)32-25(20(30)13-26)14(2)10-18-17-7-6-15-11-16(28)8-9-22(15,3)24(17,27)19(29)12-23(18,25)4/h8-9,11,14,17-19,29H,5-7,10,12-13H2,1-4H3/t14-,17-,18-,19-,22-,23-,24-,25-/m0/s1
SMILES CCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CCl
IUPAC Name [(8S,9R,10S,11S,13S,14S,16S,17R)-17-(2-chloroacetyl)-9-fluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] propanoate
Molecular Weight (g/mol) 466.97
Quantity 100 mg
Formula Weight 466.1922 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.