missing translation for 'onlineSavingsMsg'
Learn More

Clobetasone Butyrate, Mikromol™

Product Code. 18093760
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18093760 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18093760 Supplier Mikromol Supplier No. MM0207.00
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Clobetasone Butyrate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Clobetasone Butyrate
CAS 25122-57-0
Analyte or Component Names Clobetasone Butyrate
Shipping Condition Room Temperature
Molecular Formula C26 H32 Cl F O5
Certifications/Compliance ISO 17034
Synonym Clobetasone butyrate, 21-Chloro-9-fluoro-16beta-methyl-3,11,20-trioxopregna-1,4-dien-17-yl butanoate, Pregna-1,4-diene-3,11,20-trione, 21-chloro-9-fluoro-17-hydroxy-16beta-methyl-, butyrate (8CI), (16beta)-21-Chloro-9-fluoro-16-methyl-17-(1-oxobutoxy)pregna-1,4-diene-3,11,20-trione, Butyric acid, ester with 21-chloro-9-fluoro-17-hydroxy-16beta-methylpregna-1,4-diene-3,11,20-trione (8CI), CCl 5537, Clobetasone 17-butyrate, Emovate, Eumovate, GR 2/1214, Kindavate, Molivate, SN 203, CCI 5537, 2-bromoclobetasone butyrate (GW 695668X), 11-dehydrobetamethasone-17,21-dibutyrate (GW 695665X), 4,5-dihydroclobetasone butyrate (GW 695670X), 1,2-dihydroclobetasone butyrate (CCI 10702), clobetasone-17-isobutyrate (CCI 8943), 16alpha - methyl clobetasone butyrate (CCI 6999), clobetasone propionate (CCI 5473)
InChI Formula InChI=1S/C26H32ClFO5/c1-5-6-22(32)33-26(21(31)14-27)15(2)11-19-18-8-7-16-12-17(29)9-10-23(16,3)25(18,28)20(30)13-24(19,26)4/h9-10,12,15,18-19H,5-8,11,13-14H2,1-4H3/t15-,18-,19-,23-,24-,25-,26-/m0/s1
SMILES CCCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)C(=O)C[C@]12C)C(=O)CCl
IUPAC Name [(8S,9R,10S,13S,14S,16S,17R)-17-(2-chloroacetyl)-9-fluoro-10,13,16-trimethyl-3,11-dioxo-7,8,12,14,15,16-hexahydro-6H-cyclopenta[a]phenanthren-17-yl] butanoate
Molecular Weight (g/mol) 478.98
Quantity 250 mg
Formula Weight 478.1922 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.