missing translation for 'onlineSavingsMsg'
Learn More

Clopidol 100 μg/mL in Acetonitrile/Methanol, Dr. Ehrenstorfer

Product Code. 30326784
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326784 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326784 Supplier Dr. Ehrenstorfer Supplier No. DREA11687000MC100
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Clopidol
CAS 2971-90-6
Analyte or Component Names Clopidol
Shipping Condition Room Temperature
Molecular Formula C7 H7 Cl2 N O
Concentration 10 μg/mL
Certifications/Compliance ISO 17034
Synonym 2,6-Dimethyl-3,5-dichloro-4-pyridinol, 3,5-Dichloro-2,6-dimethyl-4-hydroxypyridine, 3,5-Dichloro-2,6-dimethyl-4-pyridinol, 3,5-Dichloro-2,6-dimethylpyridinol, 3,5-Dichloro-4-hydroxy-2,6-dimethylpyridine, Clopidol, Coccidiostat C, Coyden, Coyden 25, Farmcoccid, Lerbek, Methylchloropindol, Methylchlorpindol, Meticlorpindol, NSC 253479, Pharmcoccid, WR 61112
InChI Formula InChI=1S/C7H7Cl2NO/c1-3-5(8)7(11)6(9)4(2)10-3/h1-2H3,(H,10,11)
SMILES Cc1nc(C)c(Cl)c(O)c1Cl
IUPAC Name 3,5-dichloro-2,6-dimethylpyridin-4-ol
Solvent or Matrix Acetonitrile/Methanol
Molecular Weight (g/mol) 192.04
Quantity 1 mL
Formula Weight 190.9905 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.