missing translation for 'onlineSavingsMsg'
Learn More

CNOX Antibody, R&D Systems™

Product Code. p-200042772 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18464821 25 μL 25µL
18275438 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18464821 Supplier Novus Biologicals Supplier No. NBP18815225ul
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

CNOX Polyclonal specifically detects CNOX in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNOX
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias candidate growth-related and time keeping constitutive hydroquinone (NADH)oxidase, Candidate growth-related and time keeping constitutive hydroquinone [NADH]oxidase, cCNOXConstitutive Ecto-NOX, Cell proliferation-inducing gene 38 protein, cNOX, CNOXbA64J21.1, ecto-NOX disulfide-thiol exchanger 1, FLJ10094, PIG38, proliferation-inducing protein 38
Gene Symbols ENOX1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:TKDQQLQFLQQTMQGMQQQLLTIQEELNNKKSELEQAKEEQSHTQALLKVLQEQLKGTKELVETNGHSHEDSNEINVLTVALVNQDRENNIEKRS
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55068
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Isotype IgG
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.