Learn More
COL4A3 Antibody (COL4A3/9395R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-44590-100ug
427.75 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
 View more versions of this product
Product Code. 30292441
  • 518.00 € / 100µg
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
COL4A3 Monoclonal antibody specifically detects COL4A3 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
COL4A3
Monoclonal
0.2 mg/ml
10 mM PBS with 0.05% BSA
Rabbit
Protein A or G purified
RUO
1285
Store at 4⪚C.
IgG κ
Immunohistochemistry
COL4A3/9395R
Unconjugated
ATS2, ATS3, collagen alpha-3(IV) chain, collagen IV, alpha-3 polypeptide, collagen, type IV, alpha 3 (Goodpasture antigen), tumstatin
Recombinant fragment (around aa1400-1670) of human COL4A3 protein (Uniprot: P02462, P08572, P29400, P53420). Exact sequence is proprietary)
100 μg
Primary
Human
Purified