missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COL4A3 Antibody (COL4A3/9395R), Novus Biologicals™
Rabbit Monoclonal Antibody
289.00 € - 518.00 €
Specifications
| Antigen | COL4A3 |
|---|---|
| Clone | COL4A3/9395R |
| Concentration | 0.2 mg/ml |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30292441
|
Novus Biologicals
NBP3-44590-100ug |
100 μg |
518.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30302619
|
Novus Biologicals
NBP3-44590-20ug |
20 μg |
289.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
COL4A3 Monoclonal antibody specifically detects COL4A3 in Human samples. It is validated for ImmunohistochemistrySpecifications
| COL4A3 | |
| 0.2 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 1285 | |
| IgG κ | |
| Protein A or G purified |
| COL4A3/9395R | |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| ATS2, ATS3, collagen alpha-3(IV) chain, collagen IV, alpha-3 polypeptide, collagen, type IV, alpha 3 (Goodpasture antigen), tumstatin | |
| Recombinant fragment (around aa1400-1670) of human COL4A3 protein (Uniprot: P02462, P08572, P29400, P53420). Exact sequence is proprietary) | |
| Primary | |
| Store at 4⪚C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title