missing translation for 'onlineSavingsMsg'
Learn More

Collagen I/III Antibody, Novus Biologicals™

Product Code. 18056155 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mL
0.25 mL
Unit Size:
0.05mL
0.25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18056155 0.25 mL 0.25mL
18094256 0.05 mL 0.05mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18056155 Supplier Novus Biologicals Supplier No. NB10064523

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Collagen I/III Polyclonal specifically detects Collagen I/III in Bovine samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Collagen I/III
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:40
Formulation PBS, 1.0% Mannitol, 1.0% Dextran with No Preservative
Gene Accession No. P02453
Gene Alias Alpha-1 type I collagen, Collagen 1/3, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen of skin, tendon and bone, alpha-1 chain, collagen, type I, alpha 1, OI4, pro-alpha-1 collagen type 1
Gene Symbols COL1A1
Host Species Rabbit
Immunogen Bovine collagen type I and III from skin
Purification Method Affinity Purified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Cytoskeleton Markers, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 1277
Test Specificity NB100-64523 reacts with both Collagen I and III. Less than 0.5% reactivity was observed with bovine albumin and immunoglobulins. Collagen is located in the extracellular matrix of connective tissues. It is part of the interacting network of proteoglycans and proteins that provides a structural framework for both soft and calcified connective tissues.
Reconstitution Reconstitute to desired concentration.
Target Species Bovine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.