missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen VI alpha 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 554.00 €
Specifications
| Antigen | Collagen VI alpha 2 |
|---|---|
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18057213
|
Novus Biologicals
NBP2-55655 |
100 μL |
554.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18608138
|
Novus Biologicals
NBP2-55655-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Collagen VI alpha 2 Polyclonal specifically detects Collagen VI alpha 2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Collagen VI alpha 2 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| collagen alpha-2(VI) chain, collagen VI, alpha-2 polypeptide, collagen, type VI, alpha 2,10PP3610, DKFZp586E1322, FLJ46862 | |
| COL6A2 | |
| IgG | |
| Affinity Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 1292 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:KGTVHFAVVITDGHVTGSPCGGIKLQAERAREEGIRLFAVAPNQNLKEQGLRDIASTPHELYRNDYATMLPDSTEIDQDTINRIIKVMKHEAYGECYKVSC | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title