missing translation for 'onlineSavingsMsg'
Learn More

Color Coded pH Buffer Solution Tested at 20°C, Reagecon™

Use color coded pH buffers for ease of use. Reagecon™ Color Coded pH Buffer Solutions Tested at 20°C are certified and NIST traceable.

Brand:  Reagecon 1100C

Code : 23

Additional Details : Weight : 0.99630kg


 View more versions of this product

Product Code. 11404579

  • 70.95 € / 1 L
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Colored pH Buffer Solutions
  • Tested at 20°C 
  • Certified by Reagecon’s ISO/IEC 17025 AccreditedTest Method
  • NIST traceable
  • Presented in various pack sizes
Specifications

Specifications

0°C
10.00
1 L
Blue
Liquid
Characteristic
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • BUFFER SOLUTION pH 10.00 ± 0.01 @ 20°C

Signal Word
  • Danger

Hazard Category
  • Reproductive toxicity Category 1B

Hazard Statement
  • H360-May damage fertility or the unborn child.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : boric acid, disodiumsalt

SDS
Documents

Documents

Certificates
Special Offers

Special Offers