Learn More
Color Coded pH Buffer Solution Tested at 20°C, Reagecon™
Use color coded pH buffers for ease of use. Reagecon™ Color Coded pH Buffer Solutions Tested at 20°C are certified and NIST traceable.
23.25 € - 70.95 €
Product Code | Brand | Quantity | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Color | Price | Quantity & Availability | ||||
11486424
|
Reagecon
11005C |
500 mL | Blue |
25.80 €
500mL |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12808861
|
Reagecon
10405C |
500 mL | Red |
23.25 €
500mL |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12201473
|
Reagecon
10705C |
500 mL | Yellow |
40.70 €
500mL |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11404579
|
Reagecon
1100C |
1 L | Blue |
70.95 €
1 L |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11464569
|
Reagecon
1040C |
1 L | Red |
66.45 €
1 L |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11484569
|
Reagecon
1070C |
1 L | Yellow |
66.55 €
1 L |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Colored pH Buffer Solutions
- Tested at 20°C
- Certified by Reagecon’s ISO/IEC 17025 AccreditedTest Method
- NIST traceable
- Presented in various pack sizes