missing translation for 'onlineSavingsMsg'
Learn More

Color Coded pH Buffer Solution Tested at 20°C, Reagecon™

Use color coded pH buffers for ease of use. Reagecon™ Color Coded pH Buffer Solutions Tested at 20°C are certified and NIST traceable.

Brand:  Reagecon 1040C

Code : 23

Additional Details : Weight : 0.99810kg

 View more versions of this product

Product Code. 11464569

  • 66.45 € / 1 L
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Colored pH Buffer Solutions
  • Tested at 20°C 
  • Certified by Reagecon’s ISO/IEC 17025 AccreditedTest Method
  • NIST traceable
  • Presented in various pack sizes
Specifications

Specifications

0°C
4.00
1 L
Red
Liquid
Characteristic
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers