missing translation for 'onlineSavingsMsg'
Learn More

COMMD5 Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. 18657211 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18657211 0.02 mL 0.02mL
18693130 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18657211 Supplier Novus Biologicals Supplier No. NBP2923250.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

COMMD5 Polyclonal antibody specifically detects COMMD5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen COMMD5
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:5000, Immunocytochemistry/ Immunofluorescence 1:50-1:200
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias COMM domain containing 5, COMM domain-containing protein 5, FLJ13008, HCARG, HT002, Hypertension-related calcium-regulated gene protein, MGC72046
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-224 of human COMMD5 (NP_054785.2). MSAVGAATPYLHHPGDSHSGRVSFLGAQLPPEVAAMARLLGDLDRSTFRKLLKFVVSSLQGEDCREAVQRLGVSANLPEEQLGALLAGMHTLLQQALRLPPTSLKPDTFRDQLQELCIPQDLVGDLASVVFGSQRPLLDSVAQQQGAWLPHVADFRWRVDVAISTSALARSLQPSVLMQLKLSDGSAYRFEVPTAKFQELRYSVALVLKEMADLEKRCERRLQD
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 28991
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.