missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C3 Mouse anti-Human, Clone: F1-8, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46609-0.025MG
This item is not returnable.
View return policy
Description
Complement C3 Monoclonal specifically detects Complement C3 in Human samples. It is validated for ELISA, Immunoprecipitation, Immunoaffinity Purification.
Specifications
| Complement C3 | |
| Monoclonal | |
| 1 mg/ml | |
| ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500, Immunoaffinity Purification | |
| AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product | |
| Complement component 3 (C3) purified from human plasma. | |
| 0.025 mg | |
| Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience | |
| 718 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA, Immunoprecipitation, Affinity Purification | |
| F1-8 | |
| Unconjugated | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction