missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C3 Mouse anti-Human, Clone: F1-8, Novus Biologicals™
Mouse Monoclonal Antibody
237.00 €
Specifications
| Antigen | Complement C3 |
|---|---|
| Clone | F1-8 |
| Dilution | ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500, Immunoaffinity Purification |
| Applications | ELISA, Immunoprecipitation, Affinity Purification |
| Classification | Monoclonal |
Description
Complement C3 Monoclonal specifically detects Complement C3 in Human samples. It is validated for ELISA, Immunoprecipitation, Immunoaffinity Purification.Specifications
| Complement C3 | |
| ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500, Immunoaffinity Purification | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product | |
| Complement component 3 (C3) purified from human plasma. | |
| Primary | |
| Store at 4°C in the dark. |
| F1-8 | |
| ELISA, Immunoprecipitation, Affinity Purification | |
| Unconjugated | |
| Mouse | |
| Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl | |
| 718 | |
| IgG1 κ | |
| Protein A or G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title