missing translation for 'onlineSavingsMsg'
Learn More

Complement C4d Antibody (12D11), R&D Systems™

Product Code. 18203441 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18203441 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18203441 Supplier Novus Biologicals Supplier No. NBP262802
  • ONLINE EXCLUSIVE PRICE
    435.85 €
    361.20 € / 50µg
    Save 74.65 €
    17% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

ComplementC4d Polyclonal specifically detects ComplementC4d in Bovine, Human, Monkey, Pig, Primate samples. It is validated for Flow Cytometry, Immunoassay, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ComplementC4d
Applications Flow Cytometry, Immunoassay, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Clone 12D11
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay
Formulation PBS with No Preservative
Gene Symbols C4A
Host Species Mouse
Immunogen Compliment factor C4D
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 720
Target Species Bovine, Human, Monkey, Pig, Primate
Content And Storage Store at 4°C.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.