missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Factor H-related 3/CFHR3 Antibody - BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Complement Factor H-related 3/CFHR3 Polyclonal antibody specifically detects Complement Factor H-related 3/CFHR3 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | Complement Factor H-related 3/CFHR3 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:50-1:500, Immunocytochemistry/ Immunofluorescence 1:50-1:100 |
| Formulation | PBS, 50% Glycerol, pH (7.3) |
| Gene Alias | CFHL3;Complement factor H-related protein 3;DOWN16;FHR3;FHR-3;HLF4 |
| Host Species | Rabbit |
| Immunogen | CFHR3. (Uniprot# Q02985) |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?